Learn More
Sino Biological Human CD31 / PECAM1 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10148H08H100
A DNA sequence encoding the extracellular domain (Met 1-Lys 601) of human CD31 (EAW94208.1) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4132