Learn More
Sino Biological Human Endoglin / CD105 / ENG Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10149H02H20
A DNA sequence encoding the extracellular domain (Met 1-Gly 586) of human CD105 (NP_001108225.1) precursor was expressed with C-terminal fused human IgG1 Fc region.
Catalog No. 50-161-4137