Learn More
Sino Biological Endoglin / CD105 Antibody, Rabbit MAb DFS Item
Manufacturer:  Sino Biological 10149R10350
This antibody was obtained from a rabbit immunized with purified, recombinant Human Endoglin / CD105 (rh Endoglin / CD105 ; Catalog#10149-H08H; NP_001108225.1; Met1-Gly586).
Catalog No. 50-172-2013