Learn More
Sino Biological Endoglin / CD105 / ENG Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10149RP02100
Produced in rabbits immunized with purified, recombinant Human Endoglin / CD105 (rh CD105; Catalog#10149-H08H; NP_001108225.1; Met 1-Gly 586). CD105 specific IgG was purified by human CD105 affinity chromatography.
Catalog No. 50-172-2025