Learn More
Sino Biological Endoglin/CD105 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10149T26200
Produced in rabbits immunized with purified, recombinant Human Endoglin/CD105 (rh Endoglin/CD105 ; Catalog#10149-H08H; NP_001108225.1; Met1-Gly586). Endoglin/CD105 specific IgG was purified by Human Endoglin/CD105 affinity chromatography.
Catalog No. 50-172-2029