Learn More
Sino Biological Human Cadherin-6 / CDH6 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10150H08H50
A DNA sequence encoding the extracellular domain of human CDH6 (AAH00019.1) precursor (Met 1-Ala 615) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4142