Learn More
Sino Biological Sin3a Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101513T10100
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Sin3a, and purified by antigen affinity chromatography.
Catalog No. 50-172-2055