Learn More
Sino Biological SUV39H2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101528T0850
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SUV39H2, and purified by antigen affinity chromatography.
Catalog No. 50-172-2072