Learn More
Sino Biological H2AFZ Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101538T3450
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human H2AFZ, and purified by antigen affinity chromatography.
Catalog No. 50-172-2084