Learn More
Sino Biological ARID1A Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101567T32200
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human ARID1A, and purified by antigen affinity chromatography.
Catalog No. 50-172-2119