Learn More
Sino Biological Human tPA / PLAT Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10157H01H210
A DNA sequence encoding the β chain (Ile 311-Pro 562) of mature human tPA (NP_000921.1) was expressed with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-4155