Learn More
Sino Biological Human tPA / PLAT Protein DFS Item
Manufacturer:  Sino Biological 10157HNCH210
The β chain (Ile 311-Pro 562) of mature human tPA (NP_000921.1) was obtained after cleavage of the N-terminal human IgG1 Fc region from the purified chimera.
Catalog No. 50-161-4157