Learn More
Sino Biological Bad Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101576T08200
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Bad, and purified by antigen affinity chromatography.
Catalog No. 50-172-2134