Learn More
Sino Biological Human CTSZ Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10159H08H20
A DNA sequence encoding the human CTSZ (Q9UBR2) (Met 1-Val 303) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4160