Learn More
Sino Biological FACTOR D/Adipsin Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10160MM2250
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human FACTOR D/Adipsin (rh FACTOR D/Adipsin; Catalog#10160-H08H; P00746; Ile26-Ala253). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2183