Learn More
Sino Biological Human CSF1R / MCSF Receptor / CD115 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10161H08H50
A DNA sequence encoding the extracellular domain (Met 1-Glu512) of human M-CSFR (NP_005202.2) precursor was expressed with the fused C-terminal polyhistide-tagged.
Catalog No. 50-161-4168