Learn More
Sino Biological Human CSF1R / MCSF Receptor / CD115 Protein (His & GST Tag) DFS Item
Manufacturer:  Sino Biological 10161H20B150
A DNA sequence encoding the human CSF1R (NP_005202.2) cytoplasmic domain (Lys 543-Cys 972) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-4170