Learn More
Sino Biological CSF1R / MCSF Receptor / CD115 Antibody (FITC), Mouse MAb DFS Item
Manufacturer:  Sino Biological 10161MM04F25
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CSF1R / MCSF Receptor / CD115 (rh CSF1R / MCSF Receptor / CD115; Catalog#10161-H08H; NP_005202.2; Met1-Glu512) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-2205