Learn More
Sino Biological CSF1R / MCSF Receptor / CD115 Antibody, Rabbit PAb 100uL
Supplier:  Sino Biological 10161RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MCSFR / CSF1R extracellular domain (rhMCSFR; Catalog#10161-H08H; Met 1-Glu512; NP_005202.2). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-2213