Learn More
Sino Biological GPS2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101644T32200
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human GPS2, and purified by antigen affinity chromatography.
Catalog No. 50-172-2241