Learn More
Sino Biological Human IL2Ra / CD25 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10165H08H20
A DNA sequence encoding the extracellular domainof human IL2 receptor α (NP_000408.1) (Met 1-Cys 213) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4187