Learn More
Sino Biological CD25 / IL2RA Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10165MM1450
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD25 / IL2RA (rh CD25 / IL2RA; Catalog#10165-H08H; NP_000408.1; Met1-Cys213). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2274