Learn More
Sino Biological JMJD5/KDM8 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101664T4050
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human JMJD5/KDM8, and purified by antigen affinity chromatography.
Catalog No. 50-172-2318