Learn More
Sino Biological NME1/NDKA Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101691T38200
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human NME1/NDKA, and purified by antigen affinity chromatography.
Catalog No. 50-172-2359