Learn More
Sino Biological PCBP2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101698T40200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human PCBP2, and purified by antigen affinity chromatography.
Catalog No. 50-172-2371