Learn More
Sino Biological DKK1 / Dkk-1 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10170MM031
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Dkk1 (rhDkk1; Catalog#10170-H08H; NP_036374.1; Met 2-His266). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2405