Learn More
Sino Biological PIWIL4 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101709T1050
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human PIWIL4, and purified by antigen affinity chromatography.
Catalog No. 50-172-2404