Learn More
Sino Biological Human DLL4 / Delta4 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10171H08H100
A DNA sequence encoding the extracellular domain (Met 1-Pro 524) of human DLL4 (NP_061947.1) pro-protein was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4206