Learn More
Sino Biological DLL4 / Delta-4 Antibody, Rabbit MAb DFS Item
Manufacturer:  Sino Biological 10171R0031
This antibody was obtained from a rabbit immunized with purified, recombinant Human DLL4 / Delta4 extracellular domain (rh DLL4; Catalog#10171-H08H; NP_061947.1; Met 1-Pro 524).
Catalog No. 50-172-2442