Learn More
Sino Biological DLL4 / Delta-4 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10171RP01400
Produced in rabbits immunized with purified, recombinant Human DLL4 / Delta4 extracellular domain (rh DLL4; Catalog#10171-H08H; NP_061947.1; Met 1-Pro 524). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-2447