Learn More
Sino Biological PRKRA Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101724T42200
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PRKRA, and purified by antigen affinity chromatography.
Catalog No. 50-172-2455