Learn More
Sino Biological PRMT5 / SKB1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101725T08100
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human PRMT5 / SKB1, and purified by antigen affinity chromatography.
Catalog No. 50-172-2457