Learn More
Sino Biological COMP / THBS5 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10173MM0250
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human COMP (rhCOMP; Catalog#10173-H08H; NP_000086.2; Met 1-Ala 757). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2471