Learn More
Sino Biological Human Contactin 3 / CNTN3 Protein (708 Asp/Asn, His Tag) DFS Item
Manufacturer:  Sino Biological 10174H08H50
A DNA sequence encoding the human CNTN3 (NP_065923.1) (Met 1-Ser1002, 708 Asp/Asn) precursor was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4215