Learn More
Sino Biological Contactin 3 / CNTN3 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10174RP02400
Produced in rabbits immunized with purified, recombinant Human Contactin-3 (rhCNTN3; Catalog#10174-H08H; Met 1-Ser1002; NP_065923.1). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-2497