Learn More
Sino Biological Human DR6 / TNFRSF21 Protein DFS Item
Manufacturer:  Sino Biological 10175HCCH200
The mature form of human DR6 (NP_055267.1) extracellular domain (Met 1-Leu 350) with five amino acids (DDDDK) at the C-terminus was expressed and purified.
Catalog No. 50-161-4221