Learn More
Sino Biological DR6 / TNFRSF21 Antibody, Rabbit MAb DFS Item
Manufacturer:  Sino Biological 10175R00150
This antibody was obtained from a rabbit immunized with purified, recombinant Human TNFRSF21 (rh TNFRSF21; Catalog#10175-H08H; Met 1-Leu 350; NP_055267.1) .
Catalog No. 50-172-2514