Learn More
Sino Biological TNFRSF21 / DR6 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10175T4850
Produced in rabbits immunized with purified, recombinant Human TNFRSF21 / DR6 (rh TNFRSF21 / DR6; Catalog#10175-H08H; NP_055267.1; Met1-Leu350). TNFRSF21 / DR6 specific IgG was purified by Human TNFRSF21 / DR6 affinity chromatography.
Catalog No. 50-172-2520