Learn More
Sino Biological IL18RAP Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10176RP01400
Produced in rabbits immunized with purified, recombinant Human IL18RAP (rh IL18RAP; Catalog#10176-H08H; NP_003844.1; Met1-Gly357). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-2548