Learn More
Sino Biological IL18RAP Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10176T4850
Produced in rabbits immunized with purified, recombinant Human IL18RAP (rh IL18RAP; Catalog#10176-H08H; NP_003844.1; Met1-Gly357). IL18RAP specific IgG was purified by Human IL18RAP affinity chromatography.
Catalog No. 50-172-2554