Learn More
Sino Biological Human IL-1R8 / IL1RAPL1 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10177H08H10
A DNA sequence encoding the human IL1R8 (NP_055086.1) extracellular domain (Met 1-Leu 354) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4228