Learn More
Sino Biological DP2 / TFDP2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101770T40100
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human DP2 / TFDP2 , and purified by antigen affinity chromatography.
Catalog No. 50-172-2555