Learn More
Sino Biological Human Contactin 4 / CNTN4 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10178H08B20
A DNA sequence encoding the mature form of human CNTN4 isoform 1 (Q8IWV2-1) (Met 1-Ser 1000) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4230