Learn More
Sino Biological Contactin 4/CNTN4 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10178T16100
Produced in rabbits immunized with purified, recombinant Human Contactin 4/CNTN4 (rh Contactin 4/CNTN4; Catalog#10178-H08B; Q8IWV2-1; Met1-Ser1000). Contactin 4/CNTN4 specific IgG was purified by Human Contactin 4/CNTN4 affinity chromatography.
Catalog No. 50-172-2572