Learn More
Sino Biological NR1D2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101780T44100
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human NR1D2 , and purified by antigen affinity chromatography.
Catalog No. 50-172-2561