Learn More
Sino Biological NR5A1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101781T36100
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NR5A1, and purified by antigen affinity chromatography.
Catalog No. 50-172-2564