Learn More
Sino Biological Human S100A1 Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10179H01H50
A DNA sequence encoding the human S100A1 (NP_006262.1) (Gly 2-Ser94) was expressed with the N-terminal fused Fc region of human IgG1.
Catalog No. 50-161-4233