Learn More
Sino Biological S100A2 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10180MM09100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human S100A2 (rh S100A2 ; Catalog#10180-HNAE; NP_005969.1; Met2-Pro98). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2584