Learn More
Sino Biological S100A2 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10180RP01200
Produced in rabbits immunized with purified, recombinant Human S100A2 (rh S100A2 ; Catalog#10180-HNAE; NP_005969.1; Met2-Pro98). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-2588