Learn More
Sino Biological Human S100B Protein (Fc Tag) 100ug
Supplier: Sino Biological 10181H01H100
A DNA sequence encoding the human S100B (NP_006263.1) (Ser 2-Glu 92) was expressed with the fused Fc region of human IgG1 at the N-terminus.